Assay Details
| Gene Symbol: MTERF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092039.4 | XP_001092039.2 | 1157 | 115 | ||
| XM_015134126.2 | XP_014989612.1 | 1158 | 115 | |||
| GenBank mRNA | JU336419.1 | - | 1116 | 115 | ||
| JU477235.1 | - | 1094 | 115 | |||
More Information
Set Membership: |
|

MTERF1
Genomic Map