Assay Details
| Gene Symbol: TCL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144450.2 | XP_014999936.2 | 211 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015144450.2 transcripts.
Set Membership: |
|

TCL1A
Genomic Map