Assay Details
| Gene Symbol: TDRD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111794.2 | XP_014967280.2 | 381 | 92 | ||
| XM_028831945.1 | XP_028687778.1 | 381 | 92 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015111794.2, XM_028831945.1 transcripts.
Set Membership: |
|

TDRD15
Genomic Map