Assay Details
| Gene Symbol: LOC106998007 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136831.1 | XP_014992317.1 | 146 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015136831.1 transcripts.
Set Membership: |
|

LOC106998007
Genomic Map