Assay Details
| Gene Symbol: SHOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842081.1 | XP_028697914.1 | 680 | 66 | ||
| Gene Symbol: LOC719166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028843006.1 | XP_028698839.1 | 680 | 66 | ||
More Information
Set Membership: |
|

SHOX
Genomic Map