Assay Details
| Gene Symbol: TCP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137683.2 | XP_014993169.1 | 1330 | 112 | ||
| XM_015137684.2 | XP_014993170.1 | 1325 | 112 | |||
| XM_015137685.2 | XP_014993171.1 | 1344 | 112 | |||
More Information
Set Membership: |
|

TCP10
Genomic Map