Assay Details
| Gene Symbol: TCP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137683.2 | XP_014993169.1 | 106 | 98 | ||
| XM_015137688.2 | XP_014993174.1 | 105 | 98 | |||
| XM_015137689.2 | XP_014993175.2 | 106 | 98 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015137683.2, XM_015137688.2, XM_015137689.2 transcripts.
Set Membership: |
|

TCP10
Genomic Map