Assay Details
Gene Symbol: GFY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113029.4 | XP_001113029.3 | 1546 | 90 | ||
XM_015124572.2 | XP_014980058.2 | 1803 | 90 | |||
XM_028839080.1 | XP_028694913.1 | 2460 | 90 | |||
XM_028839082.1 | XP_028694915.1 | 2096 | 90 | |||
XM_028839083.1 | XP_028694916.1 | 2177 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
