Assay Details
| Gene Symbol: APOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115088.4 | XP_001115088.1 | 110 | 78 | ||
| XM_015152012.2 | XP_015007498.1 | 101 | 78 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001115088.4, XM_015152012.2 transcripts.
Set Membership: |
|

APOF
Genomic Map