Assay Details
| Gene Symbol: GPR39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116212.4 | XP_001116212.2 | 1493 | 98 | ||
| XM_028830654.1 | XP_028686487.1 | 1349 | 98 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR39| Gene Symbol: GPR39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116212.4 | XP_001116212.2 | 1493 | 98 | ||
| XM_028830654.1 | XP_028686487.1 | 1349 | 98 | |||
Set Membership: |
|