Assay Details
| Gene Symbol: PRSS47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117254.2 | XP_014972740.1 | 567 | 104 | ||
| XM_028835327.1 | XP_028691160.1 | 569 | 104 | |||
| XM_028835328.1 | XP_028691161.1 | 568 | 104 | |||
More Information
Set Membership: |
|

PRSS47
Genomic Map