Assay Details
| Gene Symbol: SULT1A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028840332.1 | XP_028696165.1 | 369 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028840332.1 transcripts.
Set Membership: |
|

SULT1A3
Genomic Map