Assay Details
| Gene Symbol: ANKRD36C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028832314.1 | XP_028688147.1 | 7042 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD36C| Gene Symbol: ANKRD36C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028832314.1 | XP_028688147.1 | 7042 | 94 | ||