Assay Details
| Gene Symbol: LOC698937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148746.2 | XP_015004232.1 | 1075 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC698937| Gene Symbol: LOC698937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148746.2 | XP_015004232.1 | 1075 | 97 | ||
Set Membership: |
|