Assay Details
| Gene Symbol: LOC100428987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133371.2 | XP_014988857.2 | 1549 | 86 | ||
| XM_028845709.1 | XP_028701542.1 | 1549 | 86 | |||
More Information
Set Membership: |
|

LOC100428987
Genomic Map