Assay Details
| Gene Symbol: LOC719317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123199.2 | XP_014978685.2 | 1342 | 102 | ||
| XM_015123200.2 | XP_014978686.2 | 1388 | 102 | |||
| XM_028840263.1 | XP_028696096.1 | 1278 | 102 | |||
| XM_028840264.1 | XP_028696097.1 | 1406 | 102 | |||
More Information
Set Membership: |
|

LOC719317
Genomic Map