Assay Details
| Gene Symbol: SPATA31D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117997.2 | XP_014973483.2 | 57 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015117997.2 transcripts.
Set Membership: |
|

SPATA31D1
Genomic Map