Assay Details
| Gene Symbol: POU4F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100319.2 | XP_001100319.1 | 305 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001100319.2 transcripts.
Set Membership: |
|

POU4F3
Genomic Map