Assay Details
| Gene Symbol: C4H6orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193881.2 | NP_001180810.2 | 70 | 84 | ||
| XM_015135469.2 | XP_014990955.2 | 254 | 84 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C4H6orf52| Gene Symbol: C4H6orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193881.2 | NP_001180810.2 | 70 | 84 | ||
| XM_015135469.2 | XP_014990955.2 | 254 | 84 | |||