Assay Details
| Gene Symbol: C4H6orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193881.2 | NP_001180810.2 | 268 | 120 | ||
| XM_015135469.2 | XP_014990955.2 | 452 | 120 | |||
| XM_015135471.2 | XP_014990957.2 | 434 | 120 | |||
| GenBank mRNA | JU485814.1 | - | 120 | |||
More Information
Set Membership: |
|

C4H6orf52
Genomic Map