Assay Details
| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194908.1 | NP_001181837.1 | 378 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194908.1 transcripts.
Set Membership: |
|

RAB9A
Genomic Map