Assay Details
| Gene Symbol: CRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266846.1 | NP_001253775.1 | 187 | 97 | ||
| XM_028850262.1 | XP_028706095.1 | 204 | 97 | |||
| GenBank mRNA | JU333005.1 | - | 181 | 97 | ||
| JU474361.1 | - | 187 | 97 | |||
More Information
Set Membership: |
|

CRIP1
Genomic Map