Assay Details
| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193753.2 | NP_001180682.1 | 719 | 87 | ||
| XM_015151080.2 | XP_015006566.1 | 962 | 87 | |||
| XM_028828815.1 | XP_028684648.1 | 616 | 87 | |||
| GenBank mRNA | JU323609.1 | - | 614 | 87 | ||
| JU471571.1 | - | 540 | 87 | |||
| JV668176.1 | - | 542 | 87 | |||
| JV717430.1 | - | 616 | 87 | |||
More Information
Set Membership: |
|

MANSC1
Genomic Map