Assay Details
| Gene Symbol: Masp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022257.1 | NP_071593.1 | 4 - 5 | 661 | 74 | |
| XM_006248526.2 | XP_006248588.1 | 4 - 5 | 905 | 74 | ||
| XM_008768804.1 | XP_008767026.1 | 4 - 5 | 899 | 74 | ||
| GenBank mRNA | AF004661.1 | - | 3 - 4 | 531 | 74 | |
| AJ277423.1 | - | 3 - 4 | 557 | 74 | ||
| AJ457084.1 | - | 4 - 5 | 661 | 74 | ||
| AJ487622.1 | - | 3 - 4 | 457 | 74 | ||
| BC085685.1 | - | 4 - 5 | 754 | 7191018 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P189476 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P163356 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P531741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45927 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P273777 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P163358 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P531741 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P189476 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P189476 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P273777 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P163358 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P531741 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P273777 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P163358 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P531741 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P189476 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P531741 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P472974 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P273777 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P189476 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P520175 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701288576 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5668 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14185 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92846 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52287 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89582 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701288576 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10751962 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751961 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751965 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751951 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751956 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751949 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751963 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751959 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751955 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751953 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751964 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751954 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751968 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751967 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751950 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751952 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751960 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751958 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751966 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10751948 | - | - |
| Rat Genome 230 2.0 Array | 1390585_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389543_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389543_at | - | - |
| Rat Genome 230 2.0 Array | 1371116_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371116_at | - | - |
| Rat Genome 230 2.0 Array | 1390585_at | - | - |
| Affymetrix HT Rat Focus Array | 1389543_at | - | - |
| Affymetrix HT Rat Focus Array | 1390585_at | - | - |
| Affymetrix HT Rat Focus Array | 1390585_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371116_at | - | - |
| Affymetrix HT Rat Focus Array | 1371116_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389543_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100004830.rn.1 | TC1100000875.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371116_PM_at | - | - |
| Rat Expression Set 230 | 1389543_at | - | - |
| Rat Expression Set 230 | 1371116_at | - | - |
| Rat Expression Set 230 | 1389543_PM_at | - | - |
| Rat Expression Set 230 | 1390585_at | - | - |
| Rat Expression Set 230 | 1390585_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751948 | - | - |
| Rat Genome U34 Set | AF004661_at | - | - |
| Rat Genome U34 Set | rc_AA801013_at | - | - |
| Rat Genome U34 Set | rc_AI169829_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21037745 | - | - |
| ABI Rat Genome Survey Microarray | 22327286 | - | - |

Masp1
Genomic Map