Assay Details
| Gene Symbol: Pik3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133399.2 | NP_596890.2 | 16 - 17 | 2491 | 76 | |
| GenBank mRNA | AF395897.1 | - | 4 - 5 | 379 | 76 | |
| CR754046.1 | - | 3 - 4 | 307 | 76 | ||
| Gene Symbol: LOC685590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773148.1 | XP_008771370.1 | 3 | 2611 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pik3ca
Genomic Map