Assay Details
| Gene Symbol: LOC103690024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763277.1 | XP_008761499.1 | 784 | 61 | ||
| Gene Symbol: Pex5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237384.2 | XP_006237446.1 | 7 - 8 | 853 | 61 | |
| XM_006237385.2 | XP_006237447.1 | 7 - 8 | 783 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103690024
Genomic Map