Assay Details
| Gene Symbol: Nnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053601.2 | NP_446053.1 | 1 - 2 | 192 | 73 | |
| XM_008762390.1 | XP_008760612.1 | 1 - 2 | 187 | 73 | ||
| GenBank mRNA | CB711943.1 | - | 1 - 2 | 128 | 73 | |
| FQ213121.1 | - | 1 - 2 | 150 | 73 | ||
| U08290.1 | - | 1 - 2 | 111 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nnat
Genomic Map