Assay Details
| Gene Symbol: Sardh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053664.2 | NP_446116.1 | 16 - 17 | 2206 | 67 | |
| XM_006233731.2 | XP_006233793.1 | 17 - 18 | 2228 | 67 | ||
| XM_006233732.2 | XP_006233794.1 | 17 - 18 | 2664 | 67 | ||
| XM_006233733.2 | XP_006233795.1 | 17 - 18 | 2230 | 67 | ||
| GenBank mRNA | AF067650.1 | - | 16 - 17 | 2241 | 67 | |
| FQ218777.1 | - | 16 - 17 | 2502 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sardh
Genomic Map