Assay Details
| Gene Symbol: Smyd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106595.1 | NP_001100065.1 | 9 - 10 | 1365 | 59 | |
| XM_006236632.2 | XP_006236694.1 | 8 - 9 | 1357 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
heart development
biological_process
positive regulation of myotube differentiation
skeletal muscle cell differentiation
positive regulation of myoblast differentiation
negative regulation of transcription, DNA-templated
molecular_function
transcription corepressor activity
metal ion binding
chromatin remodeling
heart development
biological_process
positive regulation of myotube differentiation
skeletal muscle cell differentiation
positive regulation of myoblast differentiation
negative regulation of transcription, DNA-templated
molecular_function
transcription corepressor activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P083878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127253 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083878 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083878 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P083878 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P19816 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49595 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24572 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69440 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10863109 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863106 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863110 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863105 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863114 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863108 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863111 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863107 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863112 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863113 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10863104 | - | - |
| Rat Genome 230 2.0 Array | 1397578_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380146_at | - | - |
| Rat Genome 230 2.0 Array | 1380146_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397479_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397479_at | - | - |
| Rat Genome 230 2.0 Array | 1397578_at | - | - |
| Affymetrix HT Rat Focus Array | 1397479_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397479_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400021547.rn.1 | TC0400003050.rn.1 | RTA TC |
| Rat Expression Set 230 | 1397479_at | - | - |
| Rat Expression Set 230 | 1397578_PM_at | - | - |
| Rat Expression Set 230 | 1380146_PM_at | - | - |
| Rat Expression Set 230 | 1380146_at | - | - |
| Rat Expression Set 230 | 1397479_PM_at | - | - |
| Rat Expression Set 230 | 1397578_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10863104 | - | - |
| Rat Genome U34 Set | rc_AI010736_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21881049 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364737 | - | - |

Smyd1
Genomic Map