Assay Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173349.1 | NP_001166820.1 | 3 - 4 | 1003 | 63 | |
| XM_008773419.1 | XP_008771641.1 | 3 - 4 | 1076 | 63 | ||
| GenBank mRNA | BC160840.1 | - | 3 - 4 | 864 | 7460350 | 63 |
| BC167061.1 | - | 3 - 4 | 1003 | 8364382 | 63 | |
| FQ210020.1 | - | 3 - 4 | 991 | 63 | ||
| FQ218340.1 | - | 3 - 4 | 991 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf128
Genomic Map