Assay Details
| Gene Symbol: Slc25a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014027.1 | NP_001014049.1 | 2 - 3 | 237 | 65 | |
| XM_006230557.2 | XP_006230619.1 | 3 - 4 | 294 | 65 | ||
| XM_008759993.1 | XP_008758215.1 | 2 - 3 | 261 | 65 | ||
| XM_008759994.1 | XP_008758216.1 | 2 - 3 | 267 | 65 | ||
| XM_008759995.1 | XP_008758217.1 | 2 - 3 | 219 | 65 | ||
| XM_008759996.1 | XP_008758218.1 | 2 - 3 | 822 | 65 | ||
| GenBank mRNA | BC090010.1 | - | 2 - 3 | 237 | 7324394 | 65 |
| Gene Symbol: LOC100911440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749008.3 | XP_003749056.1 | 294 | 65 | ||
| XM_008760026.1 | XP_008758248.1 | 261 | 65 | |||
| XM_008760027.1 | XP_008758249.1 | 267 | 65 | |||
| XM_008760028.1 | XP_008758250.1 | 219 | 65 | |||
| XM_008760029.1 | XP_008758251.1 | 822 | 65 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC090010.1, NM_001014027.1, XM_003749008.3, XM_006230557.2, XM_008759993.1, XM_008759994.1, XM_008759995.1, XM_008759996.1, XM_008760026.1, XM_008760027.1, XM_008760028.1, XM_008760029.1 transcripts.
Set Membership: |
|

Slc25a22
Genomic Map