Assay Details
| Gene Symbol: RGD1562084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109294.1 | NP_001102764.1 | 794 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562084| Gene Symbol: RGD1562084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109294.1 | NP_001102764.1 | 794 | 68 | ||
Set Membership: |
|