Assay Details
| Gene Symbol: Apoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271053.1 | NP_001257982.1 | 2 - 3 | 180 | 85 | |
| NM_012501.2 | NP_036633.2 | 2 - 3 | 137 | 85 | ||
| XM_008766144.1 | XP_008764366.1 | 2 - 3 | 172 | 85 | ||
| XM_008766146.1 | XP_008764368.1 | 2 - 3 | 150 | 85 | ||
| GenBank mRNA | AW917416.1 | - | 2 - 3 | 89 | 85 | |
| FQ209522.1 | - | 2 - 3 | 104 | 85 | ||
| FQ209891.1 | - | 2 - 3 | 106 | 85 | ||
| FQ210075.1 | - | 2 - 3 | 137 | 85 | ||
| FQ210397.1 | - | 2 - 3 | 104 | 85 | ||
| FQ210428.1 | - | 2 - 3 | 108 | 85 | ||
| FQ210554.1 | - | 2 - 3 | 105 | 85 | ||
| FQ210884.1 | - | 2 - 3 | 103 | 85 | ||
| FQ211165.1 | - | 2 - 3 | 104 | 85 | ||
| FQ211318.1 | - | 2 - 3 | 106 | 85 | ||
| FQ211343.1 | - | 2 - 3 | 106 | 85 | ||
| FQ211392.1 | - | 2 - 3 | 108 | 85 | ||
| FQ211416.1 | - | 2 - 3 | 109 | 85 | ||
| FQ211454.1 | - | 2 - 3 | 108 | 85 | ||
| FQ218088.1 | - | 2 - 3 | 136 | 85 | ||
| FQ218207.1 | - | 2 - 3 | 136 | 85 | ||
| FQ218211.1 | - | 2 - 3 | 180 | 85 | ||
| FQ218286.1 | - | 2 - 3 | 107 | 85 | ||
| FQ219230.1 | - | 2 - 3 | 104 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apoc3
Genomic Map