Assay Details
| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012505.2 | NP_036637.1 | 20 - 21 | 2953 | 68 | |
| GenBank mRNA | BC085764.1 | - | 20 - 21 | 2953 | 7190910 | 68 |
| M14512.1 | - | 20 - 21 | 2938 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neurotransmitter uptake
regulation of the force of heart contraction
regulation of respiratory gaseous exchange by neurological system process
potassium ion transport
sodium ion transport
cellular sodium ion homeostasis
regulation of muscle contraction
regulation of smooth muscle contraction
regulation of striated muscle contraction
metabolic process
regulation of blood pressure
adult locomotory behavior
visual learning
potassium ion import
regulation of cardiac muscle contraction by calcium ion signaling
energy coupled proton transmembrane transport, against electrochemical gradient
ATP hydrolysis coupled proton transport
regulation of vasoconstriction
cellular potassium ion homeostasis
response to nicotine
sodium ion export from cell
locomotion
negative regulation of heart contraction
negative regulation of striated muscle contraction
negative regulation of cytosolic calcium ion concentration
relaxation of cardiac muscle
cardiac muscle contraction
cellular response to mechanical stimulus
cellular response to steroid hormone stimulus
regulation of cardiac muscle cell contraction
negative regulation of calcium ion transmembrane transport
negative regulation of calcium:sodium antiporter activity
response to glycoside
sodium:potassium-exchanging ATPase activity
protein binding
ATP binding
cation-transporting ATPase activity
metal ion binding
chaperone binding
steroid hormone binding
neurotransmitter uptake
regulation of the force of heart contraction
regulation of respiratory gaseous exchange by neurological system process
potassium ion transport
sodium ion transport
cellular sodium ion homeostasis
regulation of muscle contraction
regulation of smooth muscle contraction
regulation of striated muscle contraction
metabolic process
regulation of blood pressure
adult locomotory behavior
visual learning
potassium ion import
regulation of cardiac muscle contraction by calcium ion signaling
energy coupled proton transmembrane transport, against electrochemical gradient
ATP hydrolysis coupled proton transport
regulation of vasoconstriction
cellular potassium ion homeostasis
response to nicotine
sodium ion export from cell
locomotion
negative regulation of heart contraction
negative regulation of striated muscle contraction
negative regulation of cytosolic calcium ion concentration
relaxation of cardiac muscle
cardiac muscle contraction
cellular response to mechanical stimulus
cellular response to steroid hormone stimulus
regulation of cardiac muscle cell contraction
negative regulation of calcium ion transmembrane transport
negative regulation of calcium:sodium antiporter activity
response to glycoside
sodium:potassium-exchanging ATPase activity
protein binding
ATP binding
cation-transporting ATPase activity
metal ion binding
chaperone binding
steroid hormone binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P200869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57967 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P200869 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P200869 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P200869 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P200869 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P125878 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31416 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52326 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4783 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10770013 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770001 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770003 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770017 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770000 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770018 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770015 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770002 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770007 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770009 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770010 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770020 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770019 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770012 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770021 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770008 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770011 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770004 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770016 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769999 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10770014 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10769998 | - | - |
| Rat Genome 230 2.0 Array | 1386911_at | - | - |
| Rat Genome 230 2.0 Array | 1386911_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1386911_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1386911_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300016485.rn.1 | TC1300002280.rn.1 | RTA TC |
| Rat Expression Set 230 | 1386911_PM_at | - | - |
| Rat Expression Set 230 | 1386911_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769998 | - | - |
| Rat Genome U34 Set | D90049exon#1-2_at | - | - |
| Rat Genome U34 Set | rc_AI177026_at | - | - |
| Rat Genome U34 Set | rc_AI177026_at | - | - |
| Rat Genome U34 Set | D90049exon#1-2_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21599717 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373103 | - | - |
| RatRef-12 Expression BeadChip | ILMN_54246 | - | - |

Atp1a2
Genomic Map