Assay Details
| Gene Symbol: S100g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012521.2 | NP_036653.1 | 2 - 3 | 196 | 99 | |
| GenBank mRNA | BC059153.1 | - | 2 - 3 | 200 | 6917928 | 99 |
| J02954.1 | - | 2 - 3 | 197 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

S100g
Genomic Map