Assay Details
| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012529.3 | NP_036661.3 | 738 | 59 | ||
| GenBank mRNA | BC065307.1 | - | 720 | 5598839 | 59 | |
| BC070955.1 | - | 725 | 7104205 | 59 | ||
| BC087656.1 | - | 737 | 59 | |||
| BC099814.1 | - | 722 | 7929525 | 59 | ||
| BC127477.1 | - | 728 | 8367180 | 59 | ||
| FQ216091.1 | - | 3 - 4 | 457 | 59 | ||
| M14400.1 | - | 750 | 59 | |||
| M57664.1 | - | 4 - 5 | 601 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ckb
Genomic Map