Assay Details
| Gene Symbol: Vsnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012686.2 | NP_036818.2 | 1 - 2 | 225 | 144 | |
| GenBank mRNA | BC092197.1 | - | 1 - 2 | 225 | 7313365 | 144 |
| D10666.1 | - | 1 - 2 | 233 | 144 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC092197.1, D10666.1, NM_012686.2 transcripts.
Set Membership: |
|

Vsnl1
Genomic Map