Assay Details
| Gene Symbol: Comp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012834.1 | NP_036966.1 | 10 - 11 | 1132 | 87 | |
| XM_006252822.2 | XP_006252884.1 | 7 - 8 | 1336 | 87 | ||
| GenBank mRNA | X72914.1 | - | 10 - 11 | 1132 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Comp
Genomic Map