Assay Details
| Gene Symbol: Gpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012961.1 | NP_037093.1 | 1 | 844 | 66 | |
| XM_008767077.1 | XP_008765299.1 | 2 | 1038 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr1
Genomic Map