Assay Details
| Gene Symbol: Epo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017001.1 | NP_058697.1 | 1 - 2 | 33 | 74 | |
| GenBank mRNA | D10763.1 | - | 1 - 2 | 33 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the D10763.1, NM_017001.1 transcripts.
Set Membership: |
|

Epo
Genomic Map