Assay Details
| Gene Symbol: Ppp3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017042.2 | NP_058738.1 | 11 - 12 | 1406 | 138 | |
| GenBank mRNA | D90036.1 | - | 11 - 12 | 1406 | 138 | |
| FQ214219.1 | - | 452 | 138 | |||
| M31809.1 | - | 11 - 12 | 1406 | 138 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of T cell mediated cytotoxicity
lymphangiogenesis
response to amphetamine
protein phosphorylation
protein dephosphorylation
heart development
regulation of gene expression
dephosphorylation
calcium ion regulated exocytosis
T cell differentiation
locomotion involved in locomotory behavior
calcineurin-NFAT signaling cascade
response to cytokine
social behavior
cellular response to drug
positive regulation of insulin secretion involved in cellular response to glucose stimulus
T cell homeostasis
positive regulation of transcription from RNA polymerase II promoter
muscle cell cellular homeostasis
regulation of insulin secretion
positive regulation of NFAT protein import into nucleus
protein serine/threonine phosphatase activity
calcium-dependent protein serine/threonine phosphatase activity
calcium ion binding
protein binding
calmodulin binding
drug binding
enzyme binding
protein phosphatase 2B binding
calmodulin-dependent protein phosphatase activity
protein heterodimerization activity
protein dimerization activity
negative regulation of T cell mediated cytotoxicity
lymphangiogenesis
response to amphetamine
protein phosphorylation
protein dephosphorylation
heart development
regulation of gene expression
dephosphorylation
calcium ion regulated exocytosis
T cell differentiation
locomotion involved in locomotory behavior
calcineurin-NFAT signaling cascade
response to cytokine
social behavior
cellular response to drug
positive regulation of insulin secretion involved in cellular response to glucose stimulus
T cell homeostasis
positive regulation of transcription from RNA polymerase II promoter
muscle cell cellular homeostasis
regulation of insulin secretion
positive regulation of NFAT protein import into nucleus
protein serine/threonine phosphatase activity
calcium-dependent protein serine/threonine phosphatase activity
calcium ion binding
protein binding
calmodulin binding
drug binding
enzyme binding
protein phosphatase 2B binding
calmodulin-dependent protein phosphatase activity
protein heterodimerization activity
protein dimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P15265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85683 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15265 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15265 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P15265 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15265 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700369760 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 32238 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77858 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47447 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700369760 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10779101 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779103 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779106 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779107 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779104 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10779099 | - | - |
| Rat Genome 230 2.0 Array | 1375882_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387261_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387261_at | - | - |
| Rat Genome 230 2.0 Array | 1379314_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379314_at | - | - |
| Rat Genome 230 2.0 Array | 1375882_at | - | - |
| Affymetrix HT Rat Focus Array | 1387261_at | - | - |
| Affymetrix HT Rat Focus Array | 1379314_at | - | - |
| Affymetrix HT Rat Focus Array | 1379314_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387261_PM_at | - | - |
| Rat Expression Set 230 | 1387261_PM_at | - | - |
| Rat Expression Set 230 | 1375882_at | - | - |
| Rat Expression Set 230 | 1387261_at | - | - |
| Rat Expression Set 230 | 1375882_PM_at | - | - |
| Rat Expression Set 230 | 1379314_PM_at | - | - |
| Rat Expression Set 230 | 1379314_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10779099 | - | - |
| Rat Genome U34 Set | M31809_at | - | - |
| Rat Genome U34 Set | rc_AI170309_at | - | - |
| Rat Genome U34 Set | M31809_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21892141 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351926 | - | - |
| RatRef-12 Expression BeadChip | ILMN_48514 | - | - |

Ppp3cb
Genomic Map