Assay Details
| Gene Symbol: Rgs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017214.1 | NP_058910.1 | 1 - 2 | 151 | 97 | |
| XM_008769736.1 | XP_008767958.1 | 1 - 2 | 180 | 97 | ||
| GenBank mRNA | AF117211.1 | - | 1 - 2 | 151 | 97 | |
| FQ215804.1 | - | 1 - 2 | 97 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF117211.1, NM_017214.1, XM_008769736.1 transcripts.
Set Membership: |
|

Rgs4
Genomic Map