Assay Details
| Gene Symbol: P2ry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017255.1 | NP_058951.1 | 2 - 3 | 483 | 77 | |
| XM_006229757.2 | XP_006229819.1 | 2 - 3 | 266 | 77 | ||
| GenBank mRNA | BC061754.1 | - | 2 - 3 | 486 | 5599916 | 77 |
| L46865.1 | - | 2 - 3 | 483 | 77 | ||
| U56839.1 | - | 1 - 2 | 141 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061754.1, L46865.1, NM_017255.1, U56839.1, XM_006229757.2 transcripts.
Set Membership: |
|

P2ry2
Genomic Map