Assay Details
| Gene Symbol: Gabrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017291.3 | NP_058987.2 | 2 - 3 | 288 | 61 | |
| XM_008763609.1 | XP_008761831.1 | 2 - 3 | 71 | 61 | ||
| GenBank mRNA | X95579.1 | - | 2 - 3 | 288 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008763609.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
visual perception
ion transmembrane transport
chloride transmembrane transport
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
protein binding
GABA receptor activity
protein domain specific binding
protein homodimerization activity
protein heterodimerization activity
signal transduction
visual perception
ion transmembrane transport
chloride transmembrane transport
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
protein binding
GABA receptor activity
protein domain specific binding
protein homodimerization activity
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P017148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26976 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017148 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017148 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P017148 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11898 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35943 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99812 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81211 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10868160 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868159 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868158 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868157 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868162 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868161 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868156 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868152 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868153 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868154 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868155 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10868151 | - | - |
| Rat Genome 230 2.0 Array | 1368456_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368457_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368457_at | - | - |
| Rat Genome 230 2.0 Array | 1368456_at | - | - |
| Affymetrix HT Rat Focus Array | 1368456_at | - | - |
| Affymetrix HT Rat Focus Array | 1368457_at | - | - |
| Affymetrix HT Rat Focus Array | 1368457_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368456_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500002510.rn.1 | TC0500000404.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500002509.rn.1 | TC0500000404.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500002511.rn.1 | TC0500000404.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500002508.rn.1 | TC0500000404.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500002512.rn.1 | TC0500000404.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368457_at | - | - |
| Rat Expression Set 230 | 1368456_at | - | - |
| Rat Expression Set 230 | 1368456_PM_at | - | - |
| Rat Expression Set 230 | 1368457_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10868151 | - | - |
| Rat Genome U34 Set | X95579_at | - | - |
| Rat Genome U34 Set | X95579_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20939395 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373838 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62256 | - | - |

Gabrr1
Genomic Map