Assay Details
Gene Symbol: Fam129a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022242.2 | NP_071578.2 | 8 - 9 | 1010 | 98 | |
GenBank mRNA | AB046718.2 | - | 8 - 9 | 1010 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P068222 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P068228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68716 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P068222 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P068228 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P068228 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P068222 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P068222 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P068228 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P633664 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94203 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85163 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11053 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103454 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32495 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62746 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39665 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74725 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30130 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4425 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33627 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58669 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10764667 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764663 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764670 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764677 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764662 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764674 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764669 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764671 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764665 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764666 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764668 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764676 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764673 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764675 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764672 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764664 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10764661 | - | - |
Rat Genome 230 2.0 Array | 1387454_at | - | - |
Rat Genome 230 2.0 Array | 1387454_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387454_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387454_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300003265.rn.1 | TC1300000466.rn.1 | RTA TC |
Rat Expression Set 230 | 1387454_PM_at | - | - |
Rat Expression Set 230 | 1387454_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764661 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22005887 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366304 | - | - |
RatRef-12 Expression BeadChip | ILMN_68421 | - | - |
