Assay Details
| Gene Symbol: Gpr173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022255.1 | NP_071591.1 | 1 | 1051 | 72 | |
| GenBank mRNA | AB040804.1 | - | 1 | 1051 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr173
Genomic Map