Assay Details
| Gene Symbol: Ctgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022266.2 | NP_071602.1 | 1 - 2 | 290 | 82 | |
| GenBank mRNA | AF120275.1 | - | 1 - 2 | 290 | 82 | |
| BC072503.1 | - | 1 - 2 | 280 | 7098774 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctgf
Genomic Map