Assay Details
| Gene Symbol: Ascl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022384.1 | NP_071779.1 | 1 - 2 | 1486 | 72 | |
| GenBank mRNA | X53725.1 | - | 1 - 2 | 1486 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ascl1
Genomic Map