Assay Details
| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022667.2 | NP_073158.2 | 12 - 13 | 1778 | 94 | |
| XM_006243655.1 | XP_006243717.1 | 13 - 14 | 1760 | 94 | ||
| GenBank mRNA | M64862.1 | - | 12 - 13 | 1776 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P373483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140513 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P373483 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P373483 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P373483 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P373483 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700500869 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701429254 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57389 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93809 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67235 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39899 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51999 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3153 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89398 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701429254 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700500869 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10912625 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912622 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912619 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912628 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912626 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912620 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912615 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912624 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912621 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912627 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912617 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912623 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912618 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912616 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10912614 | - | - |
| Rat Genome 230 2.0 Array | 1387492_at | - | - |
| Rat Genome 230 2.0 Array | 1372889_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372889_at | - | - |
| Rat Genome 230 2.0 Array | 1387492_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387492_at | - | - |
| Affymetrix HT Rat Focus Array | 1387492_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800010366.rn.1 | TC0800001424.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387492_PM_at | - | - |
| Rat Expression Set 230 | 1387492_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912614 | - | - |
| Rat Genome U34 Set | M64862_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20817167 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364470 | - | - |
| RatRef-12 Expression BeadChip | ILMN_60569 | - | - |

Slco2a1
Genomic Map