Assay Details
| Gene Symbol: Pcdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022868.1 | NP_074059.1 | 1 - 2 | 2333 | 72 | |
| GenBank mRNA | AB026154.1 | - | 1 - 2 | 2333 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcdh8
Genomic Map